Brady® Chemical, Biohazard, and Hazardous Material Sign: Acid, Brady Worldwide
Supplier: Brady Worldwide
131823 131824 131819 131820 131821 131822 131825 131826
10810-370EA 25.35 CAD
10810-370 10810-372 10810-418 10810-420 10810-580 10810-582 10810-584 10810-586
Brady® Chemical, Biohazard, and Hazardous Material Sign: Acid, Brady Worldwide
Signs
Indoor/outdoor safety signs from Brady® are available in a variety of materials, allowing users to choose the material and size that best fits any application.
  • Self-Sticking Polyester (B-302) is an indoor/outdoor, high performance, pressure sensitive sign that is surface-printed polyester with a polyester overlaminate
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial, and institutional environments
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial, and institutional environments
Signs that caution against an electrical hazard or identify chemicals and hazardous material storage, chemical containment areas, and acids or solvents provide the warnings and information that keep facilities and workers safe.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS). NFPA 704. OSHA 1910.1200.