HiVeg™ Wagatsuma Agar Base, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M626-500G M626-2.5KG M626-5KG
CA95022-100EA 1436.19 CAD
CA95022-100 CA89226-060 CA89226-062
HiVeg™ Wagatsuma Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Wagatsum agar base is recommended for the performance of Kanagawa test to identify virulent vibrio parahaemolyticus strains.
Order Now
SPECIFICATIONS
Environmentally Preferable