Deoxycholate Citrate Agar, HiVeg™, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV065-500G MV065-2.5KG MV065-5KG
CA61000-328EA 1017.65 CAD
CA61000-328 CA89227-344 CA89227-346
Deoxycholate Citrate Agar, HiVeg™, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Deoxycholate citrate agar is a selective medium recommended for the isolation of enteric pathogens particularly Salmonella and Shigella species.