Rotavapor® R-100 Rotary Evaporators with I-100 Controller and V-100 Pump, BUCHI
Supplier: Büchi
Rotavapor®
11100V2002 11100V2022 11100C2102 11100C2102 11100V2102 11100C2002 11100V2002 11100C2002
CA10746-742EA 14416.29 CAD
10746-742 10746-736 CA10746-742 10746-728 10746-748 CA10746-736 10746-750 CA10746-728
Rotavapor® R-100 Rotary Evaporators with I-100 Controller and V-100 Pump, BUCHI
Evaporators Rotary Evaporators
The R-100 Rotavapor® is an entry level Rotavapor®, meeting the most essential needs in evaporation
  • Economical
  • Solid
  • Expandable
  • Reliable operation
  • Flexibility
The R-100 Rotavapor® has a user-friendly manual-lift mechanism for easy positioning of the evaporation flask in to and out of the water bath, temperature ranges from 20 to 95 °C. The V-100 vacuum pump is included and is controlled by the I-100 Interface controller. The patented Combi-clip allows for easy flask handling and vapor duct removal.
The V-100 vacuum pump is included and controlled by the I-100 Interface controller.
Certifications: CE and CSA compliant.
Accessories information: The V-100 vacuum pump and I-100 interface is included. A recirculating chiller is optional and recommended for vertical condensers.
Ordering information: All accessories and connections are included for out of the box usage. Optional safety shield, >1L evaporation flasks, different joint size glassware, valves for in-house cooling and vacuum are available as accessories (sold separately). Please contact your Avantor representative for more information.