TCBS Agar, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M189-500G M189-5KG M189-2.5KG
CA95021-464EA 1097.66 CAD
CA95021-464 CA95026-832 CA89224-732
TCBS Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
TCBS agar, granulated is recommended for the selective isolation and cultivation of V. cholerae and other enteropathogenic Vibrio's causing food poisoning.