HiVeg™ Broth Base, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
MV936-500G MV936-2.5KG MV936-5KG
CA61001-472EA 1196.27 CAD
CA61001-472 CA89229-434 CA89229-436
HiVeg™ Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Potassium cyanide HiVeg™ Broth Base with KCN supplementation is used for differentiation of the members of Enterobacteriaceae on the basis of potassium cyanide tolerance.