Sodium hydroxide ≥98% (by acidimetry), pellets, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda

Management of Change (MOC) category = N

3115-01 3115-05
JT3115-1CS 514.5 CAD
JT3115-1 JT3115-5
Sodium hydroxide ≥98% (by acidimetry), pellets, BAKER ANALYZED® ACS, J.T.Baker®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NaOH) (by acidimetry) ≥98 %
Calcium (Ca) ≤0.005 %
Chloride (Cl) ≤0.005 %
Copper (Cu) ≤0.001 %
Heavy Metals (as Ag) ≤0.001 %
Insoluble Matter ≤0.003 %
Iron (Fe) ≤0.0003 %
ACS - Magnesium (Mg) ≤0.002 %
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤0.0005 %
Nitrogen Compounds (as N) ≤0.0003 %
Phosphate (PO₄) ≤0.0002 %
Potassium (K) ≤0.02 %
Sodium Carbonate (Na₂CO₃) ≤0.4 %
ACS - Sulfate (SO₄) ≤0.003 %
Meets Reagent Specifications for testing USP/NF monographs
Appearance (white hygroscopic pellets)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR