Freeze Dry Temperature Probes, Labconco®
Supplier: VWR International
76000-972 76000-974
76000-972EA 451.88 CAD
76000-972 76000-974
Freeze Dry Temperature Probes, Labconco®
Temperature Probes
Monitor sample or shelf temperature in FreeZone Tray Dryers or Heated Product Shelf Chambers.
  • Lengths for FreeZone tray dryers or heated product shelf chambers
  • Temperature is controlled and recorded
  • Works with Lyo-Works™ OS
The 28" long temperature probe is for use with FreeZone stoppering tray dryers, bulk tray dryers or small tray dryers
For use with optional stainless steel heated shelves or as a replacement part. Temperature is controlled and recorded by Lyo-Works OS on FreeZone Freeze Dryer (sold separately).
The 10.5" long Temperature Probe is for use with Heated Product Shelf Chambers.
Ordering information: FreeZone Freeze Dryer with Lyo-Works OS and FreeZone Tray Dryer or Heated Product Shelf Chamber are required (not included).