OneRack® Tube Racks, Polyoxymethylene (POM)
Supplier: Heathrow Scientific
HS27515B HS27515C HS27515D
10147-551CS 276.29 CAD
10147-551 10147-553 10147-555
OneRack® Tube Racks, Polyoxymethylene (POM)
Racks Tube Racks
OneRack® Tube Racks are tough and highly resistant to acids, bases, solvents, and heat.
  • Made out of fiberglass-reinforced polyoxymethylene (POM)
  • Have molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
  • Unique tear drop shaped openings for better retention of tubes
  • Offers the highest chemical and heat resistance (–40 to +121 °C)
  • Autoclavable
These durable tube racks are molded in a single, continuous piece so no assembly is required and there are no detachable pieces.
Delivery information: Sold in cases of 8.