Speedy Foil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60409-12 60409-13 60409-14 60409-15
103302-130EA 105.89 CAD
103302-130 103302-132 103302-514 103302-516
Speedy Foil, Electron Microscopy Sciences
Aluminium Foils
Pharmaceutical grade, pre-cut, interleaved aluminum foil squares.
  • 2×2" test tubes, vials
  • 3×3" flasks, 50 mL tubes
  • 4×4" beakers up to 500 mL
  • 5×5" beakers up to 1 L
Speedy-Foil is a fantastic time saving product that eliminates the wasteful practice of trimming pieces from over-sized rolls. Each size ships in its own ideally sized dispensing box that ensures squares remain crisp and flat.
Faster/Easier/Aseptic – Simply grab and cover! Squares are pre-cut to fit common laboratory container openings
No More Fighting – Unrolling and sizing standard aluminum foil requires scissors, contaminates foil surface areas and wastes foil
Heavy Duty – Speedy-Foil™ is 1 mil thick allowing ultra-crisp "dead fold" characteristics creating a tighter fitting contour
Keep-It-Clean – Meets FDA requirements for direct contact with pharmaceuticals; RNase and DNase free
Variety – Multiple sizes available to fit a wide range of laboratory containers