SEM Clip Specimen Mounts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75923-11 75923-23 75923-19
103301-950EA 87.31 CAD
103301-950 103302-044 103302-462
SEM Clip Specimen Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
Specially designed for easy, quick and clean mounting of any type of thin specimen on specimen stubs.
  • Available with single and multiple clips
  • Ideal for holding silicon chips, paper, wire, threads, thin films, sheet metal, and more
  • SEM clip mounts with multiple clips can hold larger samples or multiple smaller samples
These spring-loaded clips accommodate samples of up to 2 mm thickness, samples are held securely and allow for easy change of samples. Adhesives are not required, eliminating possible outgassing issues and saving time.