Heated Product Shelf Chambers, Labconco®
Supplier: VWR International
76000-932 76000-934 76000-936 76000-940 76000-942 76000-944 76000-946 76000-948 76000-950 76000-952 76000-954 76000-956 76000-958 76000-978
76000-932EA 15989.6 CAD
76000-932 76000-934 76000-936 76000-940 76000-942 76000-944 76000-946 76000-948 76000-950 76000-952 76000-954 76000-956 76000-958 76000-978
Heated Product Shelf Chambers, Labconco®
Furniture Shelves
Heated product shelves with clear chamber for drying of bulk samples or small containers.
  • 10" diameter shelves are removable
  • Shelf temperature may be set from -20 °C (-4 °F) to 60 °C (140 °F)
  • Shelves may be placed in freezer for pre-freezing
  • Shelves are height adjustable
For use with any FreeZone Freeze Dryer except Legacy models
Each Chamber includes a 12" diameter clear acrylic cylinder, vacuum release valve and 8" diameter opening to collector on bottom of chamber. Will fit 10.12" or 14.0" diameter condensers. Short Chambers include three shelves and three temperature probes. Tall Chambers include five shelves and five temperature probes.
Chambers allow for drying of bulk samples, or drying of serum bottles, vials or microplates. The 10" diameter shelves are wireless and removable so that they may be placed in a freezer for pre-freezing. Each shelf is height adjustable to accommodate a broad variety of sample sizes. Chamber connects to a FreeZone Freeze Dryer for control and recording of shelf temperature from the Lyo-Works display.
Certifications: ETL listed. CE marked.
Ordering information: FreeZone Freeze Dryer with Lyo-Works™ OS required (not included).