MacroTubes, 5 ml
Supplier: VWR International
C2500_WARDS C2500-AST_WARD C2500-B_WARDS C2500-G C2500-OB_WARDS C2500-R_WARDS C2500-Y_WARDS C2520_WARDS C2570_WARDS C2586_WARDS C2530_WARDS C2540_WARDS C2581_WARDS C2592_WARDS
470225-006CS 98.45 CAD
470225-006 470225-008 470225-010 470225-012 470225-014 470225-016 470225-018 470225-020 470225-022 470225-024 470225-026 470225-028 470225-030 470225-032
MacroTubes, 5 ml
Tubes Centrifuge Tubes
The perfect tubes for intermediate volumes.
  • Uniform wall thickness
  • Appropriate for storage from –86 to 100 °C
  • Color-code samples for easy identification
  • RNAse-, DNAse-, Pyrogen-, and Cytotoxin-free
  • Snap-cap or screw-cap available
Made of maximum clarity, low-binding virgin polypropylene, these tubes are sized to be compatible with all standard equipment.
They're a valuable resource for boiling and for sample preservation.