Polyethylene Transfer Pipettes
Supplier: VWR International
P4117-11_WARDS P4113-11_WARDS P4114-11_WARDS P4114-14_WARDS P4133-11_WARDS P4133-14_WARDS P4111-11_WARDS P4111-14_WARDS P4121-11_WARDS P4123-11_WARDS P4117-00_WARDS P4113-00_WARDS P4114-00_WARDS P4111-00_WARDS P4121-01_WARDS P4123-01_WARDS
470225-034CS 126.85 CAD
470225-034 470225-036 470225-040 470225-042 470225-044 470225-046 470225-048 470225-050 470225-052 470225-054 470225-058 470225-060 470225-062 470225-066 470225-068 470225-070
Polyethylene Transfer Pipettes
Pipettes Pasteur/Transfer Pipettes
Disposable transfer pipettes for liquid handling.
  • Transparent, non-toxic, and inert
  • Precise, uniform drop size
  • Shatter-proof and unbreakable
  • Can be used with liquid nitrogen
  • Heat sealable for liquid storage
Unbreakable, one-piece pipettes eliminate the need for rubber bulbs, broken glass, and exposure to infected material.
Ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Pipettes can also be heat sealed for airtight sample storage.
Packaging: Available packaging includes nonsterile bulk, sterile individually wrapped,More Product Information