Avantor® Apex and Apex ll, HPLC Columns
Supplier: Avantor
4M15303 4M15344 4M10310 4M15310 4M25310 FH1310-2/N
75798-642EA 2044.71 CAD
75798-642 75798-644 75798-966 75798-972 75798-974 75798-978 75798-980 75798-984 75798-986 75830-388 76398-858
Avantor® Apex and Apex ll, HPLC Columns
Chromatography Columns
Avantor® Apex and Apex ll phases are recommended for routine analyses.
  • Conventional 100 Å pore size spherical silica
  • Narrow particle distribution
  • Controlled surface area