Avantor® Apollo Phenyl, HPLC Columns
Supplier: Avantor
36538 36544 970218 36528
75797-716EA 1356.73 CAD
75797-716 75797-718 75797-720 75797-722 75830-366 76398-200 76398-202 76398-204 76398-206
Avantor® Apollo Phenyl, HPLC Columns
Chromatography Columns
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations.
  • Easy scale-up from analytical to preparative
  • Economical price
Columns are ideal for routine analyses and for educational laboratories.