Avantor® Apollo C18, HPLC Columns
Supplier: HICHROM LIMITED
36505 36515 36511 36521 970205 36537 36543 96454/N
75797-692EA 1345.42 CAD
75797-692 75797-694 75797-696 75797-698 75797-700 75797-702 75812-868 75813-094 75813-096 75813-098 75830-376 75830-686 76398-448
Avantor® Apollo C18, HPLC Columns
Chromatography Columns
Avantor® Apollo HPLC columns use high purity, base deactivated silica for powerful separations at an economical price.
  • Easy scale-up from analytical to prep
  • Extended pH stability – 1.5 to 10.5
Columns are ideal for routine analyses and for educational laboratories.