Avantor® Hichrom, Organic Acid HPLC Columns
Supplier: Avantor
9046 9048 9646 9648 28883 28884
75830-586EA 3564.45 CAD
75830-586 75830-588 75830-590 75830-592 75830-594 75830-596
Avantor® Hichrom, Organic Acid HPLC Columns
Chromatography Columns
Avantor® Hichrom organic acid columns are ion exclusion columns packed with sulphonated polystyrene-divinylbenzene.
  • Rapid analysis of organic acids and alcohols
  • pH stable polymer resin
  • Isocratic 100% aquoeus mobile phases only, no organic solvents
  • USP L17
The OA-1000 and OA-2000 columns exhibit excellent selectivity for aliphatic and aromatic acids.
The IOA-1000 and IOA-2000 columns are suitable for the separation of citric and other acids from glucose and fructose.
As with most ion exclusion columns, a column heater is necessary for normal operating procedures.