Big Trap Gas Purifier, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
1035338 1035334 1035328 1035324 1035318 1035314 1035348 1035344
75996-970EA 847.1 CAD
75996-970 75996-972 75996-974 75996-976 75996-978 75996-980 75996-982 75996-984
Big Trap Gas Purifier, Trajan Scientific and Medical
Filters Gas Filters
Big Trap Gas Purifiers are deisgned for bulk purification applications, or where several instruments are plumbed from a single source.
  • One-piece, heavy walled aluminium tube
  • 750 cm³ capacity
  • Pressure rating up to 250 psig