Gas Purifier Kit, Trajan Scientific Americas
Supplier: Trajan Scientific and Medical
1035158 1035154 1035168 1035164
75996-962EA 1059.99 CAD
75996-962 75996-964 75996-966 75996-968
Gas Purifier Kit, Trajan Scientific Americas
Filters Gas Filters
Gas purifier kits are designed to provide fast stabilization times to reduce gas consumption and provide clean gas to GC and GCMS systems.
  • Gas purifiers and mounting unit in a complete kit
  • Full removal of gas impurities for different applications
  • Improved trace analysis