Gas Purifier, Trajan Scientific Americas
Supplier: Trajan Scientific and Medical
1035220 1035230 1035250 1035210
75996-956EA 448.01 CAD
75996-956 75996-958 75996-960 75997-114
Gas Purifier, Trajan Scientific Americas
Filters Gas Filters
Gas purifiers are designed to provide fast stabilization times to reduce gas consumption and provide clean gas to GC and GCMS systems.
  • Removal of oxygen, moisture and hydrocarbons that can impact analytical results
  • Fast stabilization times
  • Improved trace analysis