SiliaBond® Tosic Acid, SCX, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R60430B-100G R60430B-10G R60430B-1KG R60430B-250G R60430B-25G R60430B-500G R60430B-50G R60530B-100G R60530B-10G R60530B-1KG R60530B-250G R60530B-25G R60530B-500G R60530B-50G R60430B-100G R60430B-10G R60430B-1KG R60430B-250G R60430B-25G R60430B-500G R60430B-50G R60530B-100G R60530B-10G R60530B-1KG R60530B-250G R60530B-25G R60530B-500G R60530B-50G
75991-686EA 2549.74 CAD
75991-686 75991-688 75991-690 75991-692 75991-694 75991-696 75991-698 75991-700 75991-702 75991-704 75991-706 75991-708 75991-710 75991-712 75991-714 75991-716 CA75991-686 CA75991-688 CA75991-690 CA75991-692 CA75991-694 CA75991-696 CA75991-698 CA75991-700 CA75991-702 CA75991-704 CA75991-706 CA75991-708 CA75991-710 CA75991-712 CA75991-714 CA75991-716
SiliaBond® Tosic Acid, SCX, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.