SiliaBond® Propylsulfonic Acid, SCX-2, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R51230B-100G R51230B-10G R51230B-1KG R51230B-250G R51230B-25G R51230B-500G R51230B-50G R51230B-5G R51430B-100G R51430B-10G R51430B-1KG R51430B-250G R51430B-25G R51430B-500G R51430B-50G R51430B-5G R51230B-100G R51230B-10G R51230B-1KG R51230B-250G R51230B-25G R51230B-500G R51230B-50G R51230B-5G R51430B-100G R51430B-10G R51430B-1KG R51430B-250G R51430B-25G R51430B-500G R51430B-50G R51430B-5G
75991-526EA 900.3 CAD
75991-526 75991-528 75991-530 75991-532 75991-534 75991-536 75991-538 75991-540 75991-542 75991-544 75991-546 75991-548 75991-550 75991-552 75991-554 75991-556 CA75991-526 CA75991-528 CA75991-530 CA75991-532 CA75991-534 CA75991-536 CA75991-538 CA75991-540 CA75991-542 CA75991-544 CA75991-546 CA75991-548 CA75991-550 CA75991-552 CA75991-554 CA75991-556
SiliaBond® Propylsulfonic Acid, SCX-2, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.