SiliaBond® Phenyl, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R34030B-100G R34030B-10G R34030B-1KG R34030B-250G R34030B-25G R34030B-500G R34030B-50G R34030B-100G R34030B-10G R34030B-1KG R34030B-250G R34030B-25G R34030B-500G R34030B-50G
75991-348EA 991.7 CAD
75991-348 75991-350 75991-352 75991-354 75991-356 75991-358 75991-360 75991-362 75991-364 75991-366 75991-368 75991-370 75991-372 75991-374 75991-376 75991-378 CA75991-348 CA75991-350 CA75991-352 CA75991-354 CA75991-356 CA75991-358 CA75991-360 CA75991-362 CA75991-364 CA75991-366 CA75991-368 CA75991-370 CA75991-372 CA75991-374 CA75991-376 CA75991-378
SiliaBond® Phenyl, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.