SiliaBond® Amine, WAX, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R52030B-100G R52030B-10G R52030B-1KG R52030B-250G R52030B-25G R52030B-500G R52030B-50G R52130B-100G R52130B-10G R52130B-1KG R52130B-250G R52130B-25G R52130B-500G R52130B-50G R52030B-100G R52030B-10G R52030B-1KG R52030B-250G R52030B-25G R52030B-500G R52030B-50G R52130B-100G R52130B-10G R52130B-1KG R52130B-250G R52130B-25G R52130B-500G R52130B-50G
75991-558EA 760.15 CAD
75991-558 75991-560 75991-562 75991-564 75991-566 75991-568 75991-570 75991-572 75991-574 75991-576 75991-578 75991-580 75991-582 75991-584 75991-586 75991-588 CA75991-558 CA75991-560 CA75991-562 CA75991-564 CA75991-566 CA75991-568 CA75991-570 CA75991-572 CA75991-574 CA75991-576 CA75991-578 CA75991-580 CA75991-582 CA75991-584 CA75991-586 CA75991-588
SiliaBond® Amine, WAX, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.