Indoor/Outdoor Industrial Vinyl Labels, Pre-Printed Spanish Headers, Brady
Supplier: Brady Worldwide
139954 139956
10018-016EA 713.61 CAD
10018-016 10018-532
Indoor/Outdoor Industrial Vinyl Labels, Pre-Printed Spanish Headers, Brady
Labels
This durable, low-shrink vinyl with ultra-aggressive adhesive is ideal for labeling pipes, equipment, shelves, and nearly any other industrial surface.
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty, and harsh environment and outdoor surfaces
  • Conforms and adheres to difficult surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes, and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents, and general industrial grime
Certifications: RoHS compliant.