Polyester (B-855), Metal Detectable Polyester (B-854) Labels, type B30, ToughWash®, Brady
Supplier: Brady Worldwide
139803 133148 133147 133146 133142 133123 133149 139804 133143 133125 133124 133120 133119 133150 133145 133144
10018-002EA 705.87 CAD
10018-002 10018-004 10018-006 10018-008 10018-010 10018-132 10018-530 10018-534 10018-536 10018-538 10018-540 10018-542 10018-544 10018-546 10018-548 10018-550 10018-552 10018-554 10018-556 10018-558
Polyester (B-855), Metal Detectable Polyester (B-854) Labels, type B30, ToughWash®, Brady
Labels
ToughWash® material polyester film with acrylic adhesive is a strong, print-on-demand label specially formulated for the Food and Beverage processing industry.
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Capable of being detected by commercially available metal detectors
  • Label construction is tough against harsh washdown environments; withstands washdowns up to 80 °C (175 °F) and pressure up to 1500 psi
Print-on-demand label stock comprised of a top-coated polyester film with an acrylic adhesive. Material (B-855) is engineered to withstand the rigorous washdown process. Material (B-854) is designed to be metal detectable, for the added security of being able to monitor the production stream for foreign debris. Ideal for safety and facility identification labeling and signage in the most aggressive environments.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.