SiliaBond® C4, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R32030B-100G R32030B-10G R32030B-1KG R32030B-250G R32030B-25G R32030B-500G R32030B-50G R32130B-100G R32130B-10G R32130B-1KG R32130B-250G R32130B-25G R32130B-500G R32130B-50G R32030B-100G R32030B-10G R32030B-1KG R32030B-250G R32030B-25G R32030B-500G R32030B-50G R32130B-100G R32130B-10G R32130B-1KG R32130B-250G R32130B-25G R32130B-500G R32130B-50G
75991-212EA 647.93 CAD
75991-212 75991-214 75991-216 75991-218 75991-220 75991-222 75991-224 75991-226 75991-228 75991-230 75991-232 75991-234 75991-236 75991-238 75991-240 75991-242 CA75991-212 CA75991-214 CA75991-216 CA75991-218 CA75991-220 CA75991-222 CA75991-224 CA75991-226 CA75991-228 CA75991-230 CA75991-232 CA75991-234 CA75991-236 CA75991-238 CA75991-240 CA75991-242
SiliaBond® C4, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.