SiliaBond® Tridecafluoro, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R63530B-100G R63530B-10G R63530B-1KG R63530B-250G R63530B-25G R63530B-500G R63530B-50G R63530B-5G R63530B-100G R63530B-10G R63530B-1KG R63530B-250G R63530B-25G R63530B-500G R63530B-50G R63530B-5G
75992-262EA 2810.28 CAD
75992-262 75992-264 75992-266 75992-268 75992-270 75992-272 75992-274 75992-276 CA75992-262 CA75992-264 CA75992-266 CA75992-268 CA75992-270 CA75992-272 CA75992-274 CA75992-276
SiliaBond® Tridecafluoro, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.