SiliaBond® TMA Acetate, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R66430B-100G R66430B-10G R66430B-1KG R66430B-250G R66430B-25G R66430B-500G R66430B-50G R66430B-100G R66430B-10G R66430B-1KG R66430B-250G R66430B-25G R66430B-500G R66430B-50G
75993-394EA 1356.45 CAD
75993-394 75993-396 75993-398 75993-400 75993-402 75993-404 75993-406 75993-408 CA75993-394 CA75993-396 CA75993-398 CA75993-400 CA75993-402 CA75993-404 CA75993-406 CA75993-408
SiliaBond® TMA Acetate, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.