SiliaBond® Piperazine, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R60030B-100G R60030B-10G R60030B-1KG R60030B-250G R60030B-25G R60030B-500G R60030B-50G R60030B-100G R60030B-10G R60030B-1KG R60030B-250G R60030B-25G R60030B-500G R60030B-50G
75991-670EA 1738.74 CAD
75991-670 75991-672 75991-674 75991-676 75991-678 75991-680 75991-682 75991-684 CA75991-670 CA75991-672 CA75991-674 CA75991-676 CA75991-678 CA75991-680 CA75991-682 CA75991-684
SiliaBond® Piperazine, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.