SiliaBond® Pentafluorophenyl, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R67530B-100G R67530B-10G R67530B-1KG R67530B-250G R67530B-25G R67530B-50G R67530B-5G R67530B-100G R67530B-10G R67530B-1KG R67530B-250G R67530B-25G R67530B-50G R67530B-5G
75992-350EA 3941.22 CAD
75992-350 75992-352 75992-354 75992-356 75992-358 75992-362 75992-364 CA75992-350 CA75992-352 CA75992-354 CA75992-356 CA75992-358 CA75992-362 CA75992-364
SiliaBond® Pentafluorophenyl, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.