SiliaBond® Guanidine, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R68230B-100G R68230B-10G R68230B-1KG R68230B-250G R68230B-25G R68230B-500G R68230B-50G R68230B-5G R68230B-100G R68230B-10G R68230B-1KG R68230B-250G R68230B-25G R68230B-500G R68230B-50G R68230B-5G
75991-002EA 1521.98 CAD
75991-002 75991-004 75991-006 75991-008 75991-010 75991-012 75991-014 75991-016 CA75991-002 CA75991-004 CA75991-006 CA75991-008 CA75991-010 CA75991-012 CA75991-014 CA75991-016
SiliaBond® Guanidine, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.