SiliaBond® Glycidoxy, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R36030B-100G R36030B-10G R36030B-1KG R36030B-250G R36030B-25G R36030B-500G R36030B-50G R36030B-5G R36030B-100G R36030B-10G R36030B-1KG R36030B-250G R36030B-25G R36030B-500G R36030B-50G R36030B-5G
75991-408EA 672.97 CAD
75991-408 75991-410 75991-412 75991-414 75991-416 75991-418 75991-420 75991-422 CA75991-408 CA75991-410 CA75991-412 CA75991-414 CA75991-416 CA75991-418 CA75991-420 CA75991-422
SiliaBond® Glycidoxy, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.