SiliaBond® Diol, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R35030B-100G R35030B-10G R35030B-1KG R35030B-250G R35030B-25G R35030B-500G R35030B-50G R35030B-100G R35030B-10G R35030B-1KG R35030B-250G R35030B-25G R35030B-500G R35030B-50G
75991-392EA 1175.79 CAD
75991-392 75991-394 75991-396 75991-398 75991-400 75991-402 75991-404 75991-406 CA75991-392 CA75991-394 CA75991-396 CA75991-398 CA75991-400 CA75991-402 CA75991-404 CA75991-406
SiliaBond® Diol, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.