SiliaBond® Dimethylamine, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R45030B-100G R45030B-10G R45030B-1KG R45030B-250G R45030B-25G R45030B-500G R45030B-50G R45030B-5G R45030B-100G R45030B-10G R45030B-1KG R45030B-250G R45030B-25G R45030B-500G R45030B-50G R45030B-5G
75991-494EA 1017.31 CAD
75991-494 75991-496 75991-498 75991-500 75991-502 75991-504 75991-506 75991-508 CA75991-494 CA75991-496 CA75991-498 CA75991-500 CA75991-502 CA75991-504 CA75991-506 CA75991-508
SiliaBond® Dimethylamine, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.