SiliaBond® Diethylamine, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R76530B-100G R76530B-10G R76530B-1KG R76530B-250G R76530B-25G R76530B-500G R76530B-50G R76530B-100G R76530B-10G R76530B-1KG R76530B-250G R76530B-25G R76530B-500G R76530B-50G
75991-074EA 1235.18 CAD
75991-074 75991-076 75991-078 75991-080 75992-438 75992-440 75992-442 75992-444 CA75991-074 CA75991-076 CA75991-078 CA75991-080 CA75992-438 CA75992-440 CA75992-442 CA75992-444
SiliaBond® Diethylamine, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.