SiliaBond® Cyanoborohydride, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R66730B-100G R66730B-10G R66730B-1KG R66730B-250G R66730B-25G R66730B-500G R66730B-50G R66730B-100G R66730B-10G R66730B-1KG R66730B-250G R66730B-25G R66730B-500G R66730B-50G
75992-334EA 1194.57 CAD
75992-334 75992-336 75992-338 75992-340 75992-342 75992-344 75992-346 75992-348 CA75992-334 CA75992-336 CA75992-338 CA75992-340 CA75992-342 CA75992-344 CA75992-346 CA75992-348
SiliaBond® Cyanoborohydride, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.
Order Now
SPECIFICATIONS
Environmentally Preferable