SiliaBond® Carboxylic Acid, WCX, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R70030B-25G R70030B-50G R70030B-250G R70030B-250G R70030B-5G R70030B-500G R70030B-1KG R70030B-50G R70030B-100G R70030B-5G R70030B-500G R70030B-10G R70030B-100G R70030B-25G R70030B-1KG R70030B-10G
CA75992-368EA 4700.68 CAD
CA75992-368 CA75992-366 75992-368 75992-366 75991-020 CA75991-020 75991-022 CA75992-372 75991-024 CA75991-022 CA75992-370 75992-372 CA75991-024 75992-370 75991-018 CA75991-018
SiliaBond® Carboxylic Acid, WCX, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.