SiliaBond® C8 Monomeric, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R30830B-100G R30830B-10G R30830B-1KG R30830B-250G R30830B-25G R30830B-500G R30830B-50G R30830B-5G R30830B-100G R30830B-10G R30830B-1KG R30830B-250G R30830B-25G R30830B-500G R30830B-50G R30830B-5G
75991-146EA 1357 CAD
75991-146 75991-148 75991-150 75991-152 75991-154 75991-156 75991-158 75991-160 CA75991-146 CA75991-148 CA75991-150 CA75991-152 CA75991-154 CA75991-156 CA75991-158 CA75991-160
SiliaBond® C8 Monomeric, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.