SiliaBond® Silver Nitrate, AgNO₃, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R23530B-1KG R23530B-100G R23530B-10G R23530B-500G R23530B-50G R23530B-1KG R23530B-250G R23530B-25G R23530B-100G R23530B-10G R23530B-500G R23530B-50G
75990-576EA 4197.89 CAD
75990-576 75990-578 75990-584 75990-598 75990-600 75991-100 75991-102 75991-104 CA75990-576 CA75990-578 CA75990-584 CA75990-598 CA75990-600 CA75991-100 CA75991-102 CA75991-104
SiliaBond® Silver Nitrate, AgNO₃, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.