SiliaBond® C18 WPD, Wide Pore, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
R33229G-100G R33229G-10G R33229G-1KG R33229G-250G R33229G-25G R33229G-500G R33229G-50G R33229G-100G R33229G-10G R33229G-1KG R33229G-250G R33229G-25G R33229G-500G R33229G-50G
75991-284EA 1187.84 CAD
75991-284 75991-286 75991-288 75991-290 75991-292 75991-294 75991-296 75991-298 CA75991-284 CA75991-286 CA75991-288 CA75991-290 CA75991-292 CA75991-294 CA75991-296 CA75991-298
SiliaBond® C18 WPD, Wide Pore, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.