Safety Separatory Funnels, Ace Glass
Supplier: Ace Glass
7247-04 7247-05 7247-10 7247-20
75877-636EA 1051.03 CAD
75877-636 75877-638 75877-640 75877-642
Safety Separatory Funnels, Ace Glass
Funnels Separating Funnels
Plastic-coated separatory funnel provides safety from common accidents such as: a broken glass drip leaving a jagged edge, weakening the vessel integrity by scratching the surface of the glass, and spills from the stopper being removed under the pressure caused by agitation of the contents.
  • Plastic-coated borosilicate glass
  • Removable PTFE drip tip
  • Pressure relief rodaviss threaded cap and stopper
Safety separatory funnel is plastic-coated to lessen the risk of common accidents involving separatory funnels. Funnel glass only has a rodaviss top, and Ace-Thred bottom, and is supplied with a PTFE stopcock plug. Complete funnel has a rodaviss capped PTFE stopper, a PTFE drip tip, and is supplied with a PTFE stopcock plug.