Scale-Up Series Bench-Top Reactor System, Ace Glass
Supplier: Ace Glass
6449-02 6449-03 6449-04 6449-05 6449-06 6449-07 6449-08 6449-09 6449-10
75877-608EA 10283.48 CAD
75877-608 75877-618 75877-620 75877-622 75877-624 75877-626 75877-628 75877-630 75877-632
Scale-Up Series Bench-Top Reactor System, Ace Glass
Laboratory Reactor Systems
Ace Glass Scale-Up Series plain bench-top reactor system includes the essential items needed to begin building a reactor for your specific application.
  • Flask made of high-quality borosilicate glass.
  • Reactors are sized for bench-top use, and their geometry translates into ACE Kilo Scale Reactors.
  • Base system includes a flask, a CAPFE o-ring, and a stand with supports.
Scale-Up Series reactors are available in 100 mL to 6000 mL capacities, and due to their geometry, the reaction can be translated into Kilo Scale reactors.
Ordering information: BaseMore Product Information