Scale-Up Series Cylindrical Flasks, Ace Glass
Supplier: Ace Glass
6447-02 6447-04 6447-06 6447-08 6447-10 6447-12 6447-14 6447-16 6447-18
75877-558EA 1996.39 CAD
75877-558 75877-560 75877-562 75877-564 75877-566 75877-568 75877-602 75877-604 75877-606
Scale-Up Series Cylindrical Flasks, Ace Glass
Flasks Round Bottom Flasks
Scale-Up Series cylindrical flasks with an Easy Drain glass valve are graduated on both sides.
  • High-quality borosilicate glass
  • Flask is graduated on both sides for either left or right-hand orientation
  • Flasks scale up from 100 mL to 6000 mL, and their geometry translates into ACE Kilo Scale flasks
Cylindrical flasks with an Easy Drain valve which is designed to prevent accidental removal when opening, and incorporates a compression tube angled side-arm for maximum unobstructed drainage. Flasks are graduated on both sides to allow for orientation inMore Product Information