Avantor® Hichrom, Anion Exclusion HPLC Columns
Supplier: HICHROM LIMITED
269006 269068 38118/S
75830-578EA 6162.68 CAD
75830-578 75830-580 76398-214
Avantor® Hichrom, Anion Exclusion HPLC Columns
Chromatography Columns
Avantor® Hichrom Anion Exclusion columns are used for the separation of organic acids and weakly ionised anions.
  • Polymer-based for broad pH stability
  • USP L22
  • Stainless steel construction
Mobile phases contain dilute mineral acids. Small amounts of acetonitrile are added to decrease retention of hydrophobic molecules.