Support Stand for CHEMRxnHUB™ Bench Top Reactors, Chemglass
Supplier: Chemglass
CG-1949-X-10 CG-1949-X-105 CG-1949-X-15
30622-564EA 8690.76 CAD
30622-564 30622-566 30622-568
Support Stand for CHEMRxnHUB™ Bench Top Reactors, Chemglass
Retort Stands
This unique stationary reactor stand is designed to fit into most benchtop hoods
Stand can hold a single reactor. The stand provides support for the reaction vessel(s), glassware, overhead stirring motor, and temperature monitor. Stand includes removable spill tray.