Safety Cutters, ZEPHER, Martor
Supplier: Martor
06(105ATGMT)
61200-334EA 165.76 CAD
61200-334
Safety Cutters, ZEPHER, Martor
Knives
Heavy-duty glass filled safety cutter for removing multiple layers of film.
  • Improved safety feature eliminates accidental cuts
This design allows the use of two hands, which provides more push power.