Buchner Funnels, Porcelain, Table Top, Chemglass
Supplier: Chemglass
CG-1889-01 CG-1889-02
89492-814EA 1177.53 CAD
89492-814 89492-816
Buchner Funnels, Porcelain, Table Top, Chemglass
Funnels Filtration Funnels Büchner Funnels
One piece table top Buchner funnels with a lower side outlet for vacuum connection.
  • Porcelain labware is widely used in the chemistry laboratory
It has excellent resistance to all reagents except hydrofluoric acid, and is highly resistant to thermal shock while having a very low coefficient of expansion
Note: Follow gradual heating/cooling rates. Avoid contact of heated ware with a cold surface. Use an oven or hot plate and an intermediate step when quicker heating/cooling rates are required. Carefully inspect labware prior to each use and do not use any product that appears defective.
The average coefficient of expansion from 20° to 200°C is 3.56 x 10-5 gradually increasing to 4.69 x 10-5 at 1000°C. To prevent thermal stress cracks on porcelain ware do not exceed a heating/cooling rate of 200°C per hour.