PBS and TBS buffers, Rockland
Supplier: Rockland Immunochemical
MB-008 MB-011 MB-012
CAMB-008EA 182 CAD
CAMB-008 CAMB-011 CAMB-012
PBS and TBS buffers, Rockland
Buffers Electrophoresis Buffers Washing Buffers
Phosphate buffered and Tris buffered saline are suitable for multiple applications including biological diluent buffer for antibodies or other biologics
Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration.
Store container at room temperature (18° to 26° C) prior to opening. If desired, the solution may be stored at 4° C or less. Some salts may precipitate out of solution at lower temperature. Allow buffer to equilibrate to room temperature (18° to 26° C) to restore solubility of some salts.